siRNA Details
| Gene Symbol: MRPL50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019051.2 | NP_061924.1 | 2 | 418 | ||
| GenBank mRNA | AK000500.1 | - | 2 | 395 | ||
| CN388719.1 | - | 2 | 428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPL50| Gene Symbol: MRPL50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019051.2 | NP_061924.1 | 2 | 418 | ||
| GenBank mRNA | AK000500.1 | - | 2 | 395 | ||
| CN388719.1 | - | 2 | 428 | |||