siRNA Details
| Gene Symbol: FAM127C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001078173.1 | NP_001071641.1 | 1 | 1120 | ||
| GenBank mRNA | AK098443.1 | - | 1 | 593 | ||
| BC048268.1 | - | 1 | 763 | |||
| BC086860.1 | - | 1 | 1088 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM127C| Gene Symbol: FAM127C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001078173.1 | NP_001071641.1 | 1 | 1120 | ||
| GenBank mRNA | AK098443.1 | - | 1 | 593 | ||
| BC048268.1 | - | 1 | 763 | |||
| BC086860.1 | - | 1 | 1088 | |||