siRNA Details
| Gene Symbol: LOC100130698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136612.1 | 6 | 970 | |||
| GenBank mRNA | AK125824.1 | - | 1 | 3201 | ||
| AK128140.1 | - | 1 | 1686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130698| Gene Symbol: LOC100130698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136612.1 | 6 | 970 | |||
| GenBank mRNA | AK125824.1 | - | 1 | 3201 | ||
| AK128140.1 | - | 1 | 1686 | |||