siRNA Details
| Gene Symbol: FAM193B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190946.2 | NP_001177875.1 | 6 | 1452 | ||
| NR_024019.3 | 9 | 3111 | ||||
| XM_006714875.2 | XP_006714938.1 | 7 | 1580 | |||
| XM_006714876.1 | XP_006714939.1 | 7 | 2303 | |||
| XM_006714878.1 | XP_006714941.1 | 8 | 2157 | |||
| XM_006714879.3 | XP_006714942.1 | 8 | 1400 | |||
| XM_006714880.1 | XP_006714943.1 | 7 | 2198 | |||
| XM_006714884.1 | XP_006714947.1 | 5 | 701 | |||
| XM_006714885.1 | XP_006714948.1 | 4 | 505 | |||
| XM_011534572.1 | XP_011532874.1 | 7 | 1475 | |||
| XM_011534573.1 | XP_011532875.1 | 8 | 1575 | |||
| XM_011534574.1 | XP_011532876.1 | 8 | 2020 | |||
| XM_011534579.2 | XP_011532881.1 | 7 | 1339 | |||
| XM_011534580.1 | XP_011532882.1 | 8 | 2364 | |||
| XM_011534581.1 | XP_011532883.1 | 7 | 2044 | |||
| XM_011534582.1 | XP_011532884.1 | 8 | 2551 | |||
| XM_011534583.1 | XP_011532885.1 | 8 | 2292 | |||
| GenBank mRNA | AB067518.1 | - | 9 | 3068 | ||
| AK001266.1 | - | 3 | 381 | |||
| AK096032.1 | - | 7 | 1263 | |||
| AL833096.1 | - | 1 | 332 | |||
| BC008784.2 | - | 5 | 701 | 3626782 | ||
| BC071696.1 | - | 3 | 355 | |||
| BC086859.1 | - | 5 | 925 | |||
| BX431446.2 | - | 3 | 655 | |||
| BX647587.1 | - | 6 | 1196 | |||

FAM193B
Genomic Map