siRNA Details
| Gene Symbol: SLC35F4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206920.1 | NP_001193849.1 | 1 | 116 | ||
| NM_001306087.1 | NP_001293016.1 | 1 | 116 | |||
| XM_017021261.1 | XP_016876750.1 | 1 | 187 | |||
| GenBank mRNA | EG328127.1 | - | 1 | 116 | ||

SLC35F4
Genomic Map