siRNA Details
| Gene Symbol: PRKXP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073405.1 | 5148 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKXP1| Gene Symbol: PRKXP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073405.1 | 5148 | ||||