siRNA Details
| Gene Symbol: NOL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198546.1 | NP_001185475.1 | 1 | 540 | ||
| NM_001198548.1 | NP_001185477.1 | 1 | 540 | |||
| NM_003787.4 | NP_003778.2 | 1 | 540 | |||
| XM_006722563.2 | XP_006722626.1 | 2 | 1297 | |||
| XM_011526237.1 | XP_011524539.1 | 2 | 1294 | |||
| XM_017026056.1 | XP_016881545.1 | 1 | 160 | |||
| XM_017026057.1 | XP_016881546.1 | 1 | 660 | |||
| XM_017026059.1 | XP_016881548.1 | 1 | 1822 | |||
| GenBank mRNA | AK297185.1 | - | 1 | 194 | ||
| BC000313.2 | - | 1 | 226 | 2821116 | ||
| DA780731.1 | - | 1 | 471 | |||

NOL4
Genomic Map