siRNA Details
| Gene Symbol: PLEKHA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019091.3 | NP_061964.3 | 2 | 530 | ||
| XM_011511689.2 | XP_011509991.1 | 2 | 592 | |||
| GenBank mRNA | AF286162.1 | - | 2 | 369 | ||
| AK074209.1 | - | 2 | 505 | |||
| AK314459.1 | - | 2 | 534 | |||
| BC044567.1 | - | 2 | 224 | 5262097 | ||
| BM794932.1 | - | 2 | 530 | |||

PLEKHA3
Genomic Map