siRNA Details
| Gene Symbol: PCDHB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019120.4 | NP_061993.3 | 1 | 1015 | ||
| GenBank mRNA | AF152501.2 | - | 1 | 770 | ||
| BC136801.1 | - | 1 | 1015 | 9020791 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDHB8| Gene Symbol: PCDHB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019120.4 | NP_061993.3 | 1 | 1015 | ||
| GenBank mRNA | AF152501.2 | - | 1 | 770 | ||
| BC136801.1 | - | 1 | 1015 | 9020791 | ||