siRNA Details
| Gene Symbol: FAM134B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034850.2 | NP_001030022.1 | 9 | 2853 | ||
| NM_019000.4 | NP_061873.2 | 7 | 2729 | |||
| XM_011514053.2 | XP_011512355.1 | 10 | 2912 | |||
| XM_011514054.2 | XP_011512356.1 | 8 | 2863 | |||
| XM_011514055.2 | XP_011512357.1 | 7 | 2653 | |||
| GenBank mRNA | AK000159.1 | - | 1 | 1773 | ||
| AK024920.1 | - | 7 | 2560 | |||
| AL832438.1 | - | 9 | 2530 | |||

FAM134B
Genomic Map