siRNA Details
Gene Symbol: LOC107986752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012904.1 | XP_016868393.1 | 1 | 3133 |
Gene Symbol: HAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X83412.1 | - | 495 |
