siRNA Details
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090593.1 | - | 3 | 279 | ||
DA025114.1 | - | 3 | 279 | |||
DA689880.1 | - | 2 | 233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090593.1 | - | 3 | 279 | ||
DA025114.1 | - | 3 | 279 | |||
DA689880.1 | - | 2 | 233 |