siRNA Details
| Gene Symbol: FAM105A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019018.2 | NP_061891.1 | 4 | 400 | ||
| GenBank mRNA | AK001989.1 | - | 4 | 359 | ||
| AK222731.1 | - | 4 | 387 | |||
| BC011524.1 | - | 4 | 326 | 3872837 | ||
| BX342178.2 | - | 4 | 407 | |||

FAM105A
Genomic Map