siRNA Details
Gene Symbol: MGC72080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002822.3 | 1, 2 | 194 | |||
GenBank mRNA | BC029615.1 | - | 1, 2 | 204 | ||
BC042035.1 | - | 1, 2 | 181 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MGC72080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002822.3 | 1, 2 | 194 | |||
GenBank mRNA | BC029615.1 | - | 1, 2 | 204 | ||
BC042035.1 | - | 1, 2 | 181 |