siRNA Details
| Gene Symbol: SLC45A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080397.2 | NP_001073866.2 | 3 | 569 | ||
| XM_011541530.1 | XP_011539832.1 | 3 | 546 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC45A1| Gene Symbol: SLC45A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080397.2 | NP_001073866.2 | 3 | 569 | ||
| XM_011541530.1 | XP_011539832.1 | 3 | 546 | |||