siRNA Details
| Gene Symbol: ANKIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019004.1 | NP_061877.1 | 3 | 652 | ||
| XM_005250458.3 | XP_005250515.1 | 3 | 670 | |||
| XM_011516334.1 | XP_011514636.1 | 3 | 670 | |||
| XM_011516335.2 | XP_011514637.1 | 3 | 234 | |||
| XM_017012359.1 | XP_016867848.1 | 3 | 670 | |||
| GenBank mRNA | AB037807.1 | - | 3 | 652 | ||
| BC063861.1 | - | 3 | 628 | |||

ANKIB1
Genomic Map