siRNA Details
| Gene Symbol: PPM1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020700.1 | NP_065751.1 | 10 | 3788 | ||
| XM_011538578.2 | XP_011536880.1 | 10 | 3924 | |||
| XM_011538579.1 | XP_011536881.1 | 10 | 3476 | |||
| GenBank mRNA | AB032983.1 | - | 9 | 3382 | ||
| AB084258.1 | - | 10 | 4053 | |||
| BC157843.1 | - | 10 | 3800 | 9057062 | ||

PPM1H
Genomic Map