siRNA Details
| Gene Symbol: GRAMD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286563.1 | NP_001273492.1 | 3, 4 | 764 | ||
| GenBank mRNA | AK127457.1 | - | 3, 4 | 764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRAMD1B| Gene Symbol: GRAMD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286563.1 | NP_001273492.1 | 3, 4 | 764 | ||
| GenBank mRNA | AK127457.1 | - | 3, 4 | 764 | ||