siRNA Details
| Gene Symbol: BAHCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291324.1 | NP_001278253.1 | 4 | 1581 | ||
| XM_011525063.2 | XP_011523365.1 | 5 | 1973 | |||
| XM_017024898.1 | XP_016880387.1 | 4 | 1632 | |||
| XM_017024899.1 | XP_016880388.1 | 4 | 1631 | |||

BAHCC1
Genomic Map