siRNA Details
| Gene Symbol: NYAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020864.1 | NP_065915.1 | 2 | 345 | ||
| XM_005246708.3 | XP_005246765.1 | 3 | 1216 | |||
| XM_011511524.1 | XP_011509826.1 | 3 | 1216 | |||
| XM_017004564.1 | XP_016860053.1 | 2 | 769 | |||
| GenBank mRNA | AB040919.1 | - | 2 | 253 | ||
| BC131727.1 | - | 2 | 275 | |||
| DA230522.1 | - | 2 | 584 | |||

NYAP2
Genomic Map