siRNA Details
| Gene Symbol: ZSWIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020883.1 | NP_065934.1 | 3 | 1313 | ||
| XM_011541861.2 | XP_011540163.1 | 3 | 1321 | |||
| XM_017001913.1 | XP_016857402.1 | 2 | 399 | |||
| GenBank mRNA | AB040944.1 | - | 3 | 1291 | ||

ZSWIM5
Genomic Map