siRNA Details
| Gene Symbol: CAMSAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080429.2 | NP_001073898.1 | 13 | 1570 | ||
| NM_020902.1 | NP_065953.1 | 11 | 1489 | |||
| XM_005272491.3 | XP_005272548.1 | 12 | 1506 | |||
| XM_011528154.2 | XP_011526456.1 | 13 | 1489 | |||
| XM_017027036.1 | XP_016882525.1 | 13 | 1543 | |||
| GenBank mRNA | AB040976.1 | - | 4 | 392 | ||
| AL833927.1 | - | 1 | 172 | |||
| BC020431.1 | - | 13 | 1413 | |||

CAMSAP3
Genomic Map