siRNA Details
Gene Symbol: PLXNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020911.1 | NP_065962.1 | 11, 12 | 2616 | ||
XM_005250686.4 | XP_005250743.1 | 11, 12 | 3014 | |||
XM_006716171.3 | XP_006716234.1 | 12, 13 | 3107 | |||
XM_011516676.2 | XP_011514978.1 | 11, 12 | 2522 | |||
XM_017012779.1 | XP_016868268.1 | 10, 11 | 2321 | |||
GenBank mRNA | AB046770.3 | - | 9, 10 | 1093 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175750, AB073228, BC141298.
Gene Ontology Categories:
