siRNA Details
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 2 | 425 | ||
XM_011516246.2 | XP_011514548.1 | 2 | 784 | |||
XM_011516247.2 | XP_011514549.1 | 2 | 785 | |||
XM_011516249.2 | XP_011514551.1 | 2 | 789 | |||
XM_011516250.2 | XP_011514552.1 | 2 | 784 | |||
XM_017012231.1 | XP_016867720.1 | 2 | 791 | |||
GenBank mRNA | BI005786.1 | - | 2 | 186 |
Gene Symbol: MUC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY307931.1 | - | 2 | 304 |
