siRNA Details
| Gene Symbol: MUC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005960.1 | NP_005951.1 | 2 | 1127 | ||
| XM_011516246.2 | XP_011514548.1 | 2 | 1486 | |||
| XM_011516247.2 | XP_011514549.1 | 2 | 1487 | |||
| XM_011516249.2 | XP_011514551.1 | 2 | 1491 | |||
| XM_011516250.2 | XP_011514552.1 | 2 | 1486 | |||
| XM_017012231.1 | XP_016867720.1 | 2 | 1493 | |||

MUC3A
Genomic Map