siRNA Details
| Gene Symbol: KDM7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030647.1 | NP_085150.1 | 7 | 927 | ||
| GenBank mRNA | AK131497.1 | - | 7 | 928 | ||
| BX649115.1 | - | 2 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KDM7A| Gene Symbol: KDM7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030647.1 | NP_085150.1 | 7 | 927 | ||
| GenBank mRNA | AK131497.1 | - | 7 | 928 | ||
| BX649115.1 | - | 2 | 194 | |||