siRNA Details
Gene Symbol: ZNRD1ASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF032109.2 | - | 3 | 2721 | ||
AF032110.1 | - | 3 | 559 | |||
BX648746.1 | - | 3 | 210 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNRD1ASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF032109.2 | - | 3 | 2721 | ||
AF032110.1 | - | 3 | 559 | |||
BX648746.1 | - | 3 | 210 |