siRNA Details
| Gene Symbol: ZNRD1ASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF032111.1 | - | 1 | 1659 | ||
| BX648746.1 | - | 3 | 2075 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRD1ASP| Gene Symbol: ZNRD1ASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF032111.1 | - | 1 | 1659 | ||
| BX648746.1 | - | 3 | 2075 | |||