siRNA Details
| Gene Symbol: ZNF594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032530.1 | NP_115919.1 | 2 | 3976 | ||
| XM_005256827.2 | XP_005256884.1 | 2 | 4289 | |||
| GenBank mRNA | AB058774.2 | - | 1 | 3961 | ||
| AK131271.1 | - | 3 | 3148 | |||
| AL833185.1 | - | 2 | 2495 | |||
| BC136604.1 | - | 1 | 3958 | 9020594 | ||

ZNF594
Genomic Map