siRNA Details
| Gene Symbol: KIAA1644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099294.1 | NP_001092764.1 | 5 | 2131 | ||
| XM_005261790.3 | XP_005261847.1 | 5 | 2229 | |||
| GenBank mRNA | AB051431.1 | - | 3 | 1797 | ||
| AK090953.1 | - | 1 | 701 | |||
| BC068280.1 | - | 1 | 687 | |||

KIAA1644
Genomic Map