siRNA Details
Gene Symbol: SHANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033517.1 | NP_277052.1 | 10 | 1256 | ||
GenBank mRNA | AB569469.1 | - | 10 | 1256 | ||
BC045765.1 | - | 1 | 189 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SHANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033517.1 | NP_277052.1 | 10 | 1256 | ||
GenBank mRNA | AB569469.1 | - | 10 | 1256 | ||
BC045765.1 | - | 1 | 189 |