siRNA Details
| Gene Symbol: UNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080419.2 | NP_001073888.2 | 16 | 3768 | ||
| XM_006722145.3 | XP_006722208.1 | 19 | 8113 | |||
| XM_006722148.2 | XP_006722211.1 | 17 | 3909 | |||
| XM_011525400.1 | XP_011523702.1 | 17 | 3837 | |||
| XM_011525401.2 | XP_011523703.1 | 11 | 2925 | |||
| XM_017025247.1 | XP_016880736.1 | 19 | 8149 | |||
| XM_017025248.1 | XP_016880737.1 | 18 | 8051 | |||
| XM_017025249.1 | XP_016880738.1 | 18 | 4194 | |||
| XM_017025250.1 | XP_016880739.1 | 17 | 4096 | |||
| GenBank mRNA | AB051540.1 | - | 16 | 3751 | ||
| BC053362.1 | - | 16 | 3751 | |||

UNK
Genomic Map