siRNA Details
| Gene Symbol: LOC645513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037630.1 | 5 | 656 | |||
| GenBank mRNA | BM544557.1 | - | 5 | 628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC645513| Gene Symbol: LOC645513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037630.1 | 5 | 656 | |||
| GenBank mRNA | BM544557.1 | - | 5 | 628 | ||