siRNA Details
| Gene Symbol: TRMT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019083.2 | NP_061956.2 | 11 | 3014 | ||
| NR_135078.1 | 11 | 2773 | ||||
| XM_005270945.3 | XP_005271002.1 | 11 | 3042 | |||
| XM_017001504.1 | XP_016856993.1 | 10 | 4666 | |||
| XM_017001505.1 | XP_016856994.1 | 10 | 4655 | |||
| GenBank mRNA | AK002081.1 | - | 11 | 3001 | ||
| BC057226.1 | - | 10 | 2581 | |||
| BC075811.1 | - | 11 | 3011 | 5104751 | ||
| CR745287.1 | - | 1 | 474 | |||

TRMT13
Genomic Map