siRNA Details
Gene Symbol: PMCHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003922.1 | 2 | 1168 | |||
GenBank mRNA | AY008412.1 | - | 2 | 1168 | ||
AY008413.1 | - | 2 | 1168 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PMCHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003922.1 | 2 | 1168 | |||
GenBank mRNA | AY008412.1 | - | 2 | 1168 | ||
AY008413.1 | - | 2 | 1168 |