siRNA Details
| Gene Symbol: CPNE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308388.1 | NP_001295317.1 | 12 | 939 | ||
| NM_153635.2 | NP_705899.2 | 12 | 920 | |||
| XM_011533386.2 | XP_011531688.1 | 11 | 1096 | |||
| XM_011533388.2 | XP_011531690.1 | 11 | 1096 | |||
| XM_011533389.2 | XP_011531691.1 | 11 | 1096 | |||
| XM_017005747.1 | XP_016861236.1 | 12 | 742 | |||
| GenBank mRNA | BC035735.1 | - | 12 | 920 | 5729888 | |
| BC130375.1 | - | 12 | 791 | 40146406 | ||

CPNE9
Genomic Map