siRNA Details
| Gene Symbol: SMAGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031628.1 | NP_001026798.1 | 3 | 307 | ||
| NM_001033873.1 | NP_001029045.1 | 3 | 296 | |||
| GenBank mRNA | AK001839.1 | - | 3 | 270 | ||
| BC001400.2 | - | 3 | 275 | |||
| BC008712.1 | - | 3 | 284 | 3546069 | ||
| BC045658.1 | - | 3 | 779 | |||
| BC120965.1 | - | 3 | 272 | 40115343 | ||
| BC120966.1 | - | 3 | 272 | 40115344 | ||
| BG740791.1 | - | 3 | 316 | |||
| CA776511.1 | - | 3 | 328 | |||

SMAGP
Genomic Map