siRNA Details
| Gene Symbol: SMAGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031628.1 | NP_001026798.1 | 4 | 836 | ||
| NM_001033873.1 | NP_001029045.1 | 4 | 825 | |||
| GenBank mRNA | AF091087.1 | - | 1 | 515 | ||
| AK001839.1 | - | 4 | 799 | |||
| BC001400.2 | - | 4 | 804 | |||
| BC003379.1 | - | 1 | 511 | |||
| BC008712.1 | - | 4 | 813 | 3546069 | ||
| BC045658.1 | - | 4 | 1308 | |||
| BC120965.1 | - | 4 | 801 | 40115343 | ||
| BC120966.1 | - | 4 | 801 | 40115344 | ||

SMAGP
Genomic Map