siRNA Details
| Gene Symbol: LINC00152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024204.1 | 4 | 568 | |||
| NR_024205.1 | 3 | 404 | ||||
| NR_024206.1 | 2 | 258 | ||||
| GenBank mRNA | AK312010.1 | - | 3 | 317 | ||
| BC009508.2 | - | 2 | 223 | |||
| BC070202.1 | - | 181 | ||||
| BM989746.1 | - | 2 | 253 | |||
| CF129206.1 | - | 2 | 296 | |||
| DW440910.1 | - | 2 | 275 | |||

LINC00152
Genomic Map