siRNA Details
| Gene Symbol: HIST3H2BB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175055.2 | NP_778225.1 | 1 | 78 | ||
| GenBank mRNA | AK091220.1 | - | 1 | 78 | ||
| BC100855.1 | - | 1 | 77 | 40002412 | ||
| BC100856.1 | - | 1 | 77 | 40002414 | ||
| BC100857.1 | - | 1 | 77 | 40002416 | ||
| BC100858.1 | - | 1 | 77 | 40002418 | ||
| BC137467.1 | - | 1 | 77 | 9021467 | ||
| BC137468.1 | - | 1 | 77 | 9021468 | ||

HIST3H2BB
Genomic Map