siRNA Details
| Gene Symbol: TM6SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001524.2 | NP_001001524.2 | 10 | 1183 | ||
| GenBank mRNA | AF255923.1 | - | 11 | 1054 | ||
| AK026307.1 | - | 7 | 1467 | |||
| AK097070.1 | - | 7 | 1529 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM6SF2| Gene Symbol: TM6SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001524.2 | NP_001001524.2 | 10 | 1183 | ||
| GenBank mRNA | AF255923.1 | - | 11 | 1054 | ||
| AK026307.1 | - | 7 | 1467 | |||
| AK097070.1 | - | 7 | 1529 | |||