siRNA Details
| Gene Symbol: FLYWCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142499.1 | NP_001135971.1 | 4 | 786 | ||
| NM_001142500.1 | NP_001135972.1 | 4 | 876 | |||
| NM_138439.2 | NP_612448.1 | 4 | 880 | |||
| XM_005255078.4 | XP_005255135.1 | 3 | 1083 | |||
| GenBank mRNA | AL542248.3 | - | 4 | 767 | ||
| BC014089.2 | - | 4 | 758 | 4330613 | ||

FLYWCH2
Genomic Map