siRNA Details
| Gene Symbol: STON1-GTF2A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198593.1 | NP_001185522.1 | 3, 4 | 2243 | ||
| NM_001198594.1 | NP_001185523.1 | 2, 3 | 2179 | |||
| NM_172311.2 | NP_758515.1 | 3, 4 | 2246 | |||
| GenBank mRNA | AF026169.1 | - | 3, 4 | 2246 | ||
| AK309740.1 | - | 3, 4 | 2243 | |||
| BC101772.1 | - | 2, 3 | 2159 | 8069278 | ||
| BC101774.1 | - | 2, 3 | 2159 | 8069280 | ||
| BC143520.1 | - | 2, 3 | 2159 | 9052029 | ||
| BX091338.1 | - | 2, 3 | 313 | |||

STON1-GTF2A1L
Genomic Map