siRNA Details
| Gene Symbol: KRTAP19-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181611.1 | NP_853642.1 | 1 | 65 | ||
| GenBank mRNA | AB096946.1 | - | 1 | 92 | ||
| BC100836.1 | - | 1 | 65 | 40002081 | ||
| BC100837.2 | - | 1 | 65 | 40002082 | ||
| BC100838.2 | - | 1 | 65 | 40002083 | ||
| BC100839.2 | - | 1 | 65 | 40002084 | ||
| BC113020.1 | - | 1 | 65 | 40013750 | ||
| BC113021.1 | - | 1 | 65 | 40013751 | ||
| BC128032.1 | - | 1 | 65 | |||

KRTAP19-5
Genomic Map