siRNA Details
| Gene Symbol: KRTAP10-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198687.2 | NP_941960.2 | 1 | 1353 | ||
| GenBank mRNA | AB076351.1 | - | 1 | 1353 | ||
| AJ566382.1 | - | 1 | 114 | |||
| BC125048.1 | - | 1 | 1342 | 40116133 | ||
| BC125049.1 | - | 1 | 1342 | 40116140 | ||

KRTAP10-4
Genomic Map