siRNA Details
| Gene Symbol: KRTAP10-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198687.2 | NP_941960.2 | 1 | 1372 | ||
| GenBank mRNA | AB076351.1 | - | 1 | 1372 | ||
| AJ566382.1 | - | 1 | 133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP10-4| Gene Symbol: KRTAP10-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198687.2 | NP_941960.2 | 1 | 1372 | ||
| GenBank mRNA | AB076351.1 | - | 1 | 1372 | ||
| AJ566382.1 | - | 1 | 133 | |||