siRNA Details
| Gene Symbol: KRTAP10-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198690.2 | NP_941963.2 | 1 | 623 | ||
| GenBank mRNA | AB076356.1 | - | 1 | 623 | ||
| BC131613.1 | - | 1 | 599 | 40115414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP10-9| Gene Symbol: KRTAP10-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198690.2 | NP_941963.2 | 1 | 623 | ||
| GenBank mRNA | AB076356.1 | - | 1 | 623 | ||
| BC131613.1 | - | 1 | 599 | 40115414 | ||