siRNA Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019116.2 | NP_061989.2 | 7 | 1471 | ||
| XM_011545894.2 | XP_011544196.1 | 6 | 1394 | |||
| GenBank mRNA | AK096189.1 | - | 3 | 552 | ||
| AK124139.1 | - | 8 | 1247 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBFD1| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019116.2 | NP_061989.2 | 7 | 1471 | ||
| XM_011545894.2 | XP_011544196.1 | 6 | 1394 | |||
| GenBank mRNA | AK096189.1 | - | 3 | 552 | ||
| AK124139.1 | - | 8 | 1247 | |||