siRNA Details
Gene Symbol: KRTAP10-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198690.2 | NP_941963.2 | 1 | 423 | ||
GenBank mRNA | AB076356.1 | - | 1 | 423 | ||
BC131613.1 | - | 1 | 399 | 40115414 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP10-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198690.2 | NP_941963.2 | 1 | 423 | ||
GenBank mRNA | AB076356.1 | - | 1 | 423 | ||
BC131613.1 | - | 1 | 399 | 40115414 |