siRNA Details
| Gene Symbol: KRTAP10-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198692.2 | NP_941965.2 | 1 | 993 | ||
| GenBank mRNA | AB076359.1 | - | 1 | 993 | ||
| AF086314.1 | - | 1 | 173 | |||
| BX103563.1 | - | 1 | 171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP10-11| Gene Symbol: KRTAP10-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198692.2 | NP_941965.2 | 1 | 993 | ||
| GenBank mRNA | AB076359.1 | - | 1 | 993 | ||
| AF086314.1 | - | 1 | 173 | |||
| BX103563.1 | - | 1 | 171 | |||